missing translation for 'onlineSavingsMsg'
Learn More

Hydroxy Melphalan (>90%), TRC

Product Code. 30389283
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Unit Size:
1mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389283 1 mg 1mg
30395033 25 mg 25mg
30399574 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30389283 Supplier Toronto Research Chemicals Supplier No. TRCH9398501MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61733-01-5
Molecular Formula C13 H19 Cl N2 O3
Molecular Weight (g/mol) 286.75
Synonym L-Phenylalanine, 4-[(2-chloroethyl)(2-hydroxyethyl)amino]-, 4-[(2-Chloroethyl)(2-hydroxyethyl)amino]-L-phenylalanine, Hydroxymelphalan, (S)-2-Amino-3-(4-((2-chloroethyl)(2-hydroxyethyl)amino)phenyl)propanoic acid, Monohydroxy Melphalan, Monohydroxymelphalan
IUPAC Name (2S)-2-amino-3-[4-[2-chloroethyl(2-hydroxyethyl)amino]phenyl]propanoic acid
SMILES N[C@@H](Cc1ccc(cc1)N(CCO)CCCl)C(=O)O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C13H19ClN2O3/c14-5-6-16(7-8-17)11-3-1-10(2-4-11)9-12(15)13(18)19/h1-4,12,17H,5-9,15H2,(H,18,19)/t12-/m0/s1
Formula Weight 286.1084
Chemical Name or Material hydroxy Melphalan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.