missing translation for 'onlineSavingsMsg'
Learn More

Hydroxy Itraconazole, MedChemExpress

Product Code. p-200101646
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
This item is not returnable. View return policy

Product Code. 16435338

Brand: Medchem Express HY12772

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Hydroxy Itraconazole (Itraconazole metabolite Hydroxy Itraconazole; R-63373) is an active metabolite of Itraconazole (ITZ), which is a triazole antifungal agent.

Hydroxy Itraconazole (Itraconazole metabolite Hydroxy Itraconazole

Specifications

Chemical Name or Material Hydroxy Itraconazole
Physical Form Solid
CAS 112559-91-8
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C35H38Cl2N8O5
Synonym Itraconazole metabolite Hydroxy Itraconazole R-63373
Solubility Information DMSO : 100 mg/mL (138.58 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C1N(C(C)C(O)C)N=CN1C2=CC=C(N3CCN(C4=CC=C(OCC5OC(CN6N=CN=C6)(C7=CC=C(Cl)C=C7Cl)OC5)C=C4)CC3)C=C2
Molecular Weight (g/mol) 721.63
Formula Weight 721.63
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.