missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Hydroxy Dynasore

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Hydroxy Dynasore is a dynamin inhibitor (IC50 values are 0.38 and 2.6 μM for dynamin 1 and 2 respectively). Inhibits dynamin-mediated endocytosis in multiple cell types and reduces synaptic vesicle endocytosis in vitro. Prevents botulinum A internalization in hippocampal neurons. Also protects against botulinum A-induced muscle paralysis in rats. Analog of Dynasore (Cat. No. 2897).
Specifications
Specifications
| Quantity | 50 mg |
| Product Type | Hydroxy Dynasore |
| Purity | >98% |
| Molecular Weight (g/mol) | 338.31 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction