missing translation for 'onlineSavingsMsg'
Learn More

Hydroxy Dimetridazole-d3, TRC

Product Code. 30138704
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138704 5 mg 5mg
17977399 10 mg 10mg
17947829 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30138704 Supplier Toronto Research Chemicals Supplier No. TRCH8252475MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1015855-78-3
Molecular Formula C5 2H3 H4 N3 O3
Molecular Weight (g/mol) 160.15
Synonym Dimetridazole-2-hydroxy D3, 1H-Imidazole-2-methanol, 1-(methyl-d3)-5-nitro- (ACI), 1-(Methyl-d3)-5-nitro-1H-imidazole-2-methanol (ACI)
IUPAC Name [5-nitro-1-(trideuteriomethyl)imidazol-2-yl]methanol
SMILES [2H]C([2H])([2H])n1c(CO)ncc1[N+](=O)[O-]

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C5H7N3O3/c1-7-4(3-9)6-2-5(7)8(10)11/h2,9H,3H2,1H3/i1D3
Formula Weight 160.0676
Percent Purity >95%
Chemical Name or Material Dimetridazole-2-hydroxy D3
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.