missing translation for 'onlineSavingsMsg'
Learn More

Hydroxy Bosentan, TRC

Product Code. 30402001
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402001 1 mg -
30404709 10 mg -
30406570 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30402001 Supplier Toronto Research Chemicals Supplier No. TRCH8302701MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 253688-60-7
Molecular Formula C27 H29 N5 O7 S
Molecular Weight (g/mol) 567.61
Synonym Benzenesulfonamide, 4-(2-hydroxy-1,1-dimethylethyl)-N-[6-(2-hydroxyethoxy)-5-(2-methoxyphenoxy)[2,2'-bipyrimidin]-4-yl]-, 4-(2-Hydroxy-1,1-dimethylethyl)-N-[6-(2-hydroxyethoxy)-5-(2-methoxyphenoxy)[2,2'-bipyrimidin]-4-yl]benzenesulfonamide, Hydroxybosentan, Ro 48-5033, Ro 48-8634
IUPAC Name N-[6-(2-hydroxyethoxy)-5-(2-methoxyphenoxy)-2-pyrimidin-2-ylpyrimidin-4-yl]-4-(1-hydroxy-2-methylpropan-2-yl)benzenesulfonamide
SMILES COc1ccccc1Oc2c(NS(=O)(=O)c3ccc(cc3)C(C)(C)CO)nc(nc2OCCO)c4ncccn4

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C27H29N5O7S/c1-27(2,17-34)18-9-11-19(12-10-18)40(35,36)32-23-22(39-21-8-5-4-7-20(21)37-3)26(38-16-15-33)31-25(30-23)24-28-13-6-14-29-24/h4-14,33-34H,15-17H2,1-3H3,(H,30,31,32)
Formula Weight 567.1788
Chemical Name or Material Hydroxy Bosentan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.