missing translation for 'onlineSavingsMsg'
Learn More

Hydroxy-2-phenylpropyl Carbamate, TRC

Product Code. 30137638
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
100mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137638 5 mg 5mg
30134466 100 mg 100mg
30003720 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137638 Supplier Toronto Research Chemicals Supplier No. TRCH9512455MG
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25451-53-0
Molecular Formula C10H13NO3
Molecular Weight (g/mol) 195.22
Synonym 2-Phenyl-1,3-propanediol 1-Carbamate, 2-Phenyl-1,3-propanediol Monocarbamate, 2-Phenyl-1,3-propanediol Monocarbamate, 2-Phenyl-1,3-propanediyl Monocarbamate, Sch 54388, W 873, USP Felbamate Related Compound A;
IUPAC Name (3-hydroxy-2-phenylpropyl) carbamate
SMILES NC(OCC(C1=CC=CC=C1)CO)=O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C10H13NO3/c11-10(13)14-7-9(6-12)8-4-2-1-3-5-8/h1-5,9,12H,6-7H2,(H2,11,13)
Formula Weight 195.0895
Chemical Name or Material Hydroxy-2-phenylpropyl Carbamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.