missing translation for 'onlineSavingsMsg'
Learn More

(Hydroxy-2-naphthylmethyl)phosphonic acid, 98%, Thermo Scientific Chemicals

Product Code. 15446069
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15446069 5 mg 5mg
15456069 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15446069 Supplier Thermo Scientific Alfa Aesar Supplier No. J63913.#0
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A selective insulin receptor tyrosine kinase inhibitor

A selective insulin receptor tyrosine kinase inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132541-52-7
Molecular Formula C11H11O4P
Molecular Weight (g/mol) 238.179
MDL Number MFCD00674174
InChI Key AMJJLDJPDLKNJA-UHFFFAOYSA-N
Synonym hnmpa, hydroxy-2-naphthalenylmethyl phosphonic acid, hydroxy 2-naphthyl methanephosphonic acid, hydroxy naphthalen-2-yl methylphosphonic acid, hydroxy naphthalen-2-yl methyl phosphonic acid, phosphonic acid, p-hydroxy-2-naphthalenylmethyl, oh-name-pa, acmc-1cglf, biomolki_000021, biomolki2_000010
PubChem CID 129317
IUPAC Name [hydroxy(naphthalen-2-yl)methyl]phosphonic acid
SMILES C1=CC=C2C=C(C=CC2=C1)C(O)P(=O)(O)O

Specifications

Color White
Quantity 5 mg
Formula Weight 238.18
Percent Purity 98%
Physical Form Solid
Chemical Name or Material (Hydroxy-2-naphthylmethyl)phosphonic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.