missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxy(2-naphthyl)methanephosphonic Acid, TRC
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Specifications
Specifications
| Chemical Name or Material | Hydroxy(2-naphthyl)methanephosphonic Acid |
| CAS | 132541-52-7 |
| Molecular Formula | C11H11O4P |
| Quantity | 1 mg |
| Synonym | P-(Hydroxy-2-naphthalenylmethyl)phosphonic Acid, (Hydroxy-2-naphthalenylmethyl)phosphonic Acid, HNMPA |
| InChI Formula | InChI=1S/C11H11O4P/c12-11(16(13,14)15)10-6-5-8-3-1-2-4-9(8)7-10/h1-7,11-12H,(H2,13,14,15) |
| SMILES | OC(c1ccc2ccccc2c1)P(=O)(O)O |
| IUPAC Name | [hydroxy(naphthalen-2-yl)methyl]phosphonic acid |
| Molecular Weight (g/mol) | 238.18 |
| Formula Weight | 238.0395 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction