missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxocobalamin Acetate, TRC
Specifications
Specifications
| Chemical Name or Material | Hydroxocobalamin Acetate |
| CAS | 22465-48-1 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C64 H91 Co N13 O16 P |
| Quantity | 50 mg |
| Synonym | Hydroxocobalamin acetate, Coα-[α-(5,6-dimethylbenzimidazolyl)]-Coβ-hydroxocobamide acetate, Cobinamide, Co-(acetato-κO)-, dihydrogen phosphate (ester), inner salt, 3'-ester with (5,6-dimethyl-1-α-D-ribofuranosyl-1H-benzimidazole-κN3), Cobalamin, acetato- (6CI), Cobinamide, acetate (salt) hydroxide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-α-D-ribofuranosyl-1H-benzimidazole, Cobinamide, acetate (salt) hydroxide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-α-D-ribofuranosylbenzimidazole (8CI), Cobinamide, acetate phosphate, 3'-ester with 5,6-dimethyl-1-α-D-ribofuranosylbenzimidazole, inner salt (7CI), Acetatocobalamin, Depo-gamma, Fresmin S, Hydroxycobalamine acetate, Novidroxin, Twelvmin |
| InChI Formula | InChI=1S/C62H90N13O14P.C2H4O2.Co/c1-29-20-39-40(21-30(29)2)75(28-70-39)57-52(84)53(41(27-76)87-57)89-90(85,86)88-31(3)26-69-49(83)18-19-59(8)37(22-46(66)80)56-62(11)61(10,25-48(68)82)36(14-17-45(65)79)51(74-62)33(5)55-60(9,24-47(67)81)34(12-15-43(63)77)38(71-55)23-42-58(6,7)35(13-16-44(64)78)50(72-42)32(4)54(59)73-56;1-2(3)4;/h20-21,23,28,31,34-37,41,52-53,56-57,76,84H,12-19,22,24-27H2,1-11H3,(H15,63,64,65,66,67,68,69,71,72,73,74,77,78,79,80,81,82,83,85,86);1H3,(H,3,4);/q;;+3/p-3/t31-,34-,35-,36-,37+,41-,52-,53-,56-,57+,59-,60+,61+,62+;;/m1../s1 |
| SMILES | C[C@@H]1CNC(=O)CC[C@]2(C)[C@@H](CC(=O)N)[C@H]3N4C2=C(C)C5=N6C(=CC7=N8C(=C(C)C9=N([C@]3(C)[C@@](C)(CC(=O)N)[C@@H]9CCC(=O)N)[Co+]468(OC(=O)C)N%10=CN([C@H]%11O[C@H](CO)[C@@H](OP(=O)([O-])O1)[C@H]%11O)c%12cc(C)c(C)cc%10%12)[C@@](C)(CC(=O)N)[C@@H]7CCC(=O)N)C(C)(C)[C@@H]5CCC(=O)N |
| Molecular Weight (g/mol) | 1388.39 |
| Formula Weight | 1387.5776 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?