missing translation for 'onlineSavingsMsg'
Learn More

Hydroquinone, TRC

Product Code. 17940965
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17940965 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17940965 Supplier Toronto Research Chemicals Supplier No. TRCH7147155G
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 123-31-9
Molecular Formula C6 H6 O2
Quantity 5 g
Synonym Hydroquinone, 1,4-Benzoquinol, 1,4-Dihydroxybenzene, 1,4-Phenylenediol, 1,4-p-Benzenediol, 4-Hydroxyphenol, Aida, Arctuvin, BQ(H), Benzohydroquinone, Benzoquinol, Black & White Bleaching Cream, Diak 5, Dihydroquinone, Eldopacque, Eldopaque, Eldopaque Forte, Eldoquin, Eldoquin Forte, HE 5, Hydroquinol, NSC 9247, Phiaquin, Quinol, Solaquin Forte, Solution Q, Tecquinol, Tenox HQ, p-Benzenediol, p-Dihydroquinone, p-Dihydroxybenzene, p-Dioxybenzene, p-Hydroquinone, p-Hydroxyphenol, p-Phenylenediol, p-Quin
InChI Formula InChI=1S/C6H6O2/c7-5-1-2-6(8)4-3-5/h1-4,7-8H
SMILES Oc1ccc(O)cc1
IUPAC Name benzene-1,4-diol
Molecular Weight (g/mol) 110.11
Formula Weight 110.0368
Percent Purity >95%
Chemical Name or Material Hydroquinone
Grade HPLC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.