missing translation for 'onlineSavingsMsg'
Learn More

Hydroquinone sulfonic acid, potassium salt, tech., ACROS Organics™

Product Code. 10021841 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10021841 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10021841 Supplier Thermo Scientific Acros Supplier No. 354680050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 21799-87-1
Color White to Gray
Infrared Spectrum Authentic
Packaging Glass Bottle
Molecular Formula C6H5KO5S
Linear Formula (HO)2C6H3SO3K
MDL Number MFCD00007475
Quantity 5 g
Synonym potassium 2,5-dihydroxybenzenesulfonate, hydroquinonesulfonic acid potassium salt, potassium2,5-dihydroxybenzenesulfonate, unii-249e3f00ep, benzenesulfonic acid, 2,5-dihydroxy-, monopotassium salt, 2,5-dihydroxybenzenesulfonic acid potassium salt, potassium hydroquinonesulfonate, hydroquinone sulfonic acid potassium salt, hydroquinone sulfonic acid, potassium salt, benzenesulfonic acid, 2,5-dihydroxy-, potassium salt 1:1
InChI Key VKDSBABHIXQFKH-UHFFFAOYSA-M
SMILES C1=CC(=C(C=C1O)S(=O)(=O)[O-])O.[K+]
IUPAC Name potassium;2,5-dihydroxybenzenesulfonate
Molecular Weight (g/mol) 228.259
PubChem CID 23672329
Formula Weight 228.25
Grade Technical
Physical Form Powder
Chemical Name or Material Hydroquinone sulfonic acid, potassium salt, tech.
Show More Show Less
compliance-icons
Product Identifier
  • Hydroquinone sulfonic acid, potassium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.