missing translation for 'onlineSavingsMsg'
Learn More

Hydroquinone-O,O'-diacetic acid, 98%

Product Code. 11465257
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11465257

Brand: Thermo Scientific Alfa Aesar B22645.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2245-53-6 | C10H10O6 | 226.18 g/mol

Hydroquinone-O,O'-diacetic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hydroquinone-O,O′-diacetic acid is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2245-53-6
Molecular Formula C10H10O6
Molecular Weight (g/mol) 226.18
MDL Number MFCD00016816
InChI Key DNXOCFKTVLHUMU-UHFFFAOYSA-N
Synonym 2,2'-1,4-phenylenebis oxy diacetic acid, 1,4-phenylenedioxydiacetic acid, hydroquinone-o,o'-diacetic acid, 1,4-dicarboxymethoxybenzene, hydroquinone-o,o-diacetic acid, acetic acid, p-phenylenedioxy di, acetic acid, 2,2'-1,4-phenylenebis oxy bis, 2-4-carboxymethoxy phenoxy acetic acid
PubChem CID 75251
IUPAC Name 2-[4-(carboxymethoxy)phenoxy]acetic acid
SMILES OC(=O)COC1=CC=C(OCC(O)=O)C=C1

Specifications

Melting Point ∼255°C (decomposition)
Quantity 25 g
Beilstein 1984454
Solubility Information Slightly soluble in water.
Formula Weight 226.19
Percent Purity 98%
Chemical Name or Material Hydroquinone-O,O'-diacetic acid
compliance-icons
Product Identifier
  • Hydroquinone-O,O'-diacetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Hydroquinone-O,O'-diacetic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.