missing translation for 'onlineSavingsMsg'
Learn More

Hydroquinone-O,O'-diacetic acid, 98%

Product Code. 11455257
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11455257 5 g 5g
11465257 25 g 25g
11475257 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11455257 Supplier Thermo Scientific Alfa Aesar Supplier No. B22645.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 2245-53-6 | C10H10O6 | 226.18 g/mol

Hydroquinone-O,O'-diacetic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hydroquinone-O,O′-diacetic acid is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2245-53-6
Molecular Formula C10H10O6
Molecular Weight (g/mol) 226.18
MDL Number MFCD00016816
InChI Key DNXOCFKTVLHUMU-UHFFFAOYSA-N
Synonym 2,2'-1,4-phenylenebis oxy diacetic acid, 1,4-phenylenedioxydiacetic acid, hydroquinone-o,o'-diacetic acid, 1,4-dicarboxymethoxybenzene, hydroquinone-o,o-diacetic acid, acetic acid, p-phenylenedioxy di, acetic acid, 2,2'-1,4-phenylenebis oxy bis, 2-4-carboxymethoxy phenoxy acetic acid
PubChem CID 75251
IUPAC Name 2-[4-(carboxymethoxy)phenoxy]acetic acid
SMILES OC(=O)COC1=CC=C(OCC(O)=O)C=C1

Specifications

Melting Point ∼255°C (decomposition)
Quantity 5 g
Beilstein 1984454
Solubility Information Slightly soluble in water.
Formula Weight 226.19
Percent Purity 98%
Chemical Name or Material Hydroquinone-O,O'-diacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.