missing translation for 'onlineSavingsMsg'
Learn More

Hydroquinone-d4, TRC

Product Code. 30138267
Change view
Click to view available options
Quantity:
25 mg
50 mg
500 mg
Unit Size:
25mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138267 50 mg 50mg
30134837 500 mg 500mg
17909249 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30138267 Supplier Toronto Research Chemicals Supplier No. TRCH71471750MG
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25294-85-3
Molecular Formula C6H2D4O2
Molecular Weight (g/mol) 114.14
Synonym Hydroquinone (ring D4), 1,4-Benzene-2,3,5,6-d4-diol, Hydroquinone-2,3,5,6-d4 (8CI), (2H4)Benzene-1,4-diol, 1,4-Benzenediol-d4, 1,4-Benzoquinol-d4, 1,4-Dihydroxybenzene-d4, Dihydroquinone-d4, Eldopacque-d4, Eldopaque-d4, Eldopaque Forte-d4, Eldoquin-d4, 4-Hydroxyphenol-d4, p-Hydroxyphenol-d4, p-Phenylenediol-d4, p-Quinol-d4;
IUPAC Name 2,3,5,6-tetradeuteriobenzene-1,4-diol
SMILES [2H]c1c([2H])c(O)c([2H])c([2H])c1O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C6H6O2/c7-5-1-2-6(8)4-3-5/h1-4,7-8H/i1D,2D,3D,4D
Formula Weight 114.0619
Chemical Name or Material Hydroquinone-d4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.