missing translation for 'onlineSavingsMsg'
Learn More

Hydroquinone D4 (ring D4), Dr. Ehrenstorfer

Product Code. 30322150
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30322150 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30322150 Supplier Dr. Ehrenstorfer Supplier No. DREC14223010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hydroquinone-d4
CAS 25294-85-3
Molecular Formula C6H2D4O2
Quantity 50 mg
Synonym 1,4-Benzenediol-d4, 1,4-Benzoquinol-d4, 1,4-Dihydroxybenzene-d4, Dihydroquinone-d4, Eldopacque-d4, Eldopaque-d4, Eldopaque Forte-d4, Eldoquin-d4, 4-Hydroxyphenol-d4, p-Hydroxyphenol-d4, p-Phenylenediol-d4, p-Quinol-d4;
SMILES OC1=C([2H])C([2H])=C(O)C([2H])=C1[2H]
Molecular Weight (g/mol) 114.14
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.