missing translation for 'onlineSavingsMsg'
Learn More

Hydroquinone bis(2-hydroxyethyl) ether, 95%

Product Code. 11416956
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11416956 100 g 100g
11426956 500 g 500g
11436956 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11416956 Supplier Thermo Scientific Alfa Aesar Supplier No. B24037.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 104-38-1 | C10H14O4 | 198.218 g/mol

Hydroquinone bis(2-hydroxyethyl) ether is used to produce 1,4-bis-(2-bromo-ethoxy)-benzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hydroquinone bis(2-hydroxyethyl) ether is used to produce 1,4-bis-(2-bromo-ethoxy)-benzene.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Store under dry inert gas. Protect from humidity and water. It is hygroscopic in nature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-38-1
Molecular Formula C10H14O4
Molecular Weight (g/mol) 198.218
MDL Number MFCD00002861
InChI Key WTPYFJNYAMXZJG-UHFFFAOYSA-N
Synonym hydroquinone bis 2-hydroxyethyl ether, 1,4-bis 2-hydroxyethoxy benzene, vernatzer 30/10, 2,2'-1,4-phenylenebis oxy diethanol, hydroquinone di 2-hydroxyethyl ether, hydroquinone diethylol ether, 2,2'-phenylenedioxy diethanol, 2,2'-p-phenylenedioxy diethanol, 2,2'-p-phenylenedioxydiethanol, 2-4-2-hydroxyethoxy phenoxy ethanol
PubChem CID 66912
IUPAC Name 2-[4-(2-hydroxyethoxy)phenoxy]ethanol
SMILES C1=CC(=CC=C1OCCO)OCCO

Specifications

Melting Point 100°C to 102°C
Quantity 100 g
Sensitivity Hygroscopic
Solubility Information Insoluble in water.
Formula Weight 198.22
Percent Purity 95%
Chemical Name or Material Hydroquinone bis(2-hydroxyethyl) ether

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.