missing translation for 'onlineSavingsMsg'
Learn More

Hydroquinone, 99%

Codice prodotto. 11400253
Cambia vista
Click to view available options
Quantity:
250 g
1000 g
5000 g
Dimensione della confezione:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity unitSize
11400253 5000 g 5000g
11490243 250 g 250g
11480243 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 11400253 Fornitore Thermo Scientific Alfa Aesar N. del fornitore A11411.0I
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 123-31-9 | C6H6O2 | 110.112 g/mol

Hydroquinone is used as a reducing agent and finds applications in black and white photographic developers for film and paper along with 4-(methylamino)phenol sulfate (metol). In this application, silver halides are reduced to elemental silver. It acts as a polymerization inhibitor which prevents polymerization of acrylic acid, methyl methacrylate, cyanoacrylate and other monomers. It undergoes mild oxidation to convert into parabenzoquinone. It is also used as an intermediate to produce antioxidants for rubber and food.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hydroquinone is used as a reducing agent and finds applications in black and white photographic developers for film and paper along with 4-(methylamino)phenol sulfate (metol). In this application, silver halides are reduced to elemental silver. It acts as a polymerization inhibitor which prevents polymerization of acrylic acid, methyl methacrylate, cyanoacrylate and other monomers. It undergoes mild oxidation to convert into parabenzoquinone. It is also used as an intermediate to produce antioxidants for rubber and food.

Solubility
Soluble in water, methanol, ethanol, diethyl ether, acetone, benzene and carbon tetrachloride.

Notes
Air and light sensitive. Incompatible with strong bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-31-9
Molecular Formula C6H6O2
Molecular Weight (g/mol) 110.112
MDL Number MFCD00002339
InChI Key QIGBRXMKCJKVMJ-UHFFFAOYSA-N
Synonym hydroquinone, 1,4-benzenediol, quinol, 1,4-dihydroxybenzene, p-benzenediol, 4-hydroxyphenol, p-hydroquinone, p-hydroxyphenol, p-dihydroxybenzene, benzoquinol
PubChem CID 785
ChEBI CHEBI:17594
IUPAC Name benzene-1,4-diol
SMILES C1=CC(=CC=C1O)O

Specifications

Melting Point 170°C to 172°C
Density 1.32
pH 3.75
Boiling Point 285°C to 287°C
Flash Point 165°C (329°F)
Odor Odorless
Quantity 5000 g
UN Number UN3077
Beilstein 605970
Sensitivity Air and light sensitive
Merck Index 14,4808
Solubility Information Soluble in water,methanol,ethanol,diethyl ether,acetone,benzene and carbon tetrachloride.
Formula Weight 110.11
Percent Purity 99%
Chemical Name or Material Hydroquinone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.