missing translation for 'onlineSavingsMsg'
Learn More

Hydroquinine, MedChemExpress

Product Code. 16467826
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16467826 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16467826 Supplier Medchem Express Supplier No. HY42034

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Hydroquinine is a cinchona alkaloid, can be used in the preparation of its derivatives such as C9 epihydroquinine, 9-acetoxy-10,11-dihydroquinine and 10,11-dihydroquinine monohydrochloride.

Hydroquinine is a cinchona alkaloid, can be used in the preparation of its derivatives such as C9 epihydroquinine, 9-acetoxy-10,11-dihydroquinine and 10,11-dihydroquinine monohydrochloride.

Specifications

Chemical Name or Material Hydroquinine
Physical Form Solid
CAS 522-66-7
Color White to Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C20H26N2O2
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 130 mg/mL (398.25 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES CC[C@@H]1[C@@H]2C[C@H]([N@](C1)CC2)[C@H](O)C3=CC=NC4=CC=C(OC)C=C43
Molecular Weight (g/mol) 326.43
Formula Weight 326.43
Percent Purity 97.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.