missing translation for 'onlineSavingsMsg'
Learn More

Hydroquinine, 95%

Product Code. 15773447 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15773447 5 g 5g
15763447 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15773447 Supplier Thermo Scientific Acros Supplier No. 461650050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 522-66-7 | C20H26N2O2 | 326.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 522-66-7
Molecular Formula C20H26N2O2
Molecular Weight (g/mol) 326.44
MDL Number MFCD00151107
InChI Key LJOQGZACKSYWCH-ADMHDHIONA-N
Synonym dihydroquinine, hydroquinine, 1r-2s,4s,5r-5-ethylquinuclidin-2-yl 6-methoxyquinolin-4-yl methanol, unii-31j3q51t6l, hydrochinin, quinine, 10,11-dihydro, --dihydroquinine, --dihydroquinine, analytical standard, 8a,9r-10,11-dihydro-6'-methoxycinchonan-9-ol
PubChem CID 121515
IUPAC Name (R)-[(2S,4S,5R)-5-ethyl-1-azabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol
SMILES CCC1CN2CCC1CC2[C@H](O)C1=CC=NC2=CC=C(OC)C=C12

Specifications

Melting Point 168.0°C
Boiling Point 468.4°C
Flash Point 225°C
Assay Percent Range 94% min. (HPLC)
Quantity 5 g
Formula Weight 326.43
Percent Purity 95%
Chemical Name or Material Hydroquinine
compliance-icons
Product Identifier
  • Hydroquinine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Hydroquinine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.