Learn More
Brannan™ Low Tension Specific Gravity Hydrometer
The specific gravity 60/60°F scale compares the relative density of a given liquid with that of water, at 60°F (15.6°C).
Brand:  Brannan™ 10/922/0
Code : 33
Additional Details : Weight : 0.20000kg
Product Code. 12891458
  • £19.20 / Each
Estimated Shipment: 23-05-2024
to see stock.
Add to basket
Description
Description
The specific gravity 60/60°F scale compares the relative density of a given liquid with that of water, at 60°F (15.6°C).
  • Manufactured from soda glass, steel shot ballast and synthetic wax resin
  • Range of 0.050 S60/60°F
  • M50, 270mm long
  • Low tension 0.90 to 0.95
Specifications
Specifications
0.001 S60/60°F
0.001 S60/60°F
0.050 S60/60°F
Comparing relative density of a liquid to water
270 mm
Soda glass, steel shot ballast, synthetic wax resin
Steel Shot