missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell™ Hydrograin Grain Cowhide Leather Gloves
Click to view available options
Size:
8
9
11
Unit Size:
Each
Description
- Fully water-repellent grain cowhide leather
- For handling operations in a damp or greasy environment
- Ideal for indoor and outdoor applications
- Elasticated wrist
- Gunn cut
- Wing thumb
Specifications
Specifications
| Certifications/Compliance | CE CATEGORY II Intermediary risks, EN 420-2003 EEC Basic standards, EN 388-2003 Mechanical hazards, EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Certificate Number N°0075/797/162/11/06/0317, EC Attestation Number N°0075/797/162/11/06/0317 |
| Certifications/Compliance (Europe) | EN 420-2003 EEC Basic Standards, EN 388-2016 Mechanical Hazards |
| Description | Water repellent |
| Color | Yellow |
| Hand | Specific |
| Material | Grain Cowhide Leather |
| Product Type | Gloves |
| Size | 9 |
| Style | Gunn |
| For Use With (Application) | Automotive, Building, Construction, Forestry, Forklift Operation, Green Spaces, Logistics and Maintenance Industries |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction