missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Hydrogold Column Guard

Product Code. 15560312 Shop All Thermo Scientific Products
Click to view available options
Length (Metric):
2 m
5 m
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15560312

Brand: Thermo Scientific™ 26H500553

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Provides protection to the analytical column.

Protects against column contamination caused by non-volatile materials, extending the column lifetime.
TRUSTED_SUSTAINABILITY

Specifications

Type GC Guard Column
Diameter (Metric) Inner 0.53 mm
Length (Metric) 5 m
Quantity 1
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.