missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen tetranitratoaurate(III) hydrate, Premion™, 99.99% (metals basis), Au 38.9-40.1%, Thermo Scientific Chemicals

Product Code. 11394028
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11394028 1 g 1g
11384028 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11394028 Supplier Thermo Scientific Alfa Aesar Supplier No. 040431.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used for Isomerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00046178
Synonym Nitratoauric acid

Specifications

Melting Point ∼72°C (decomposition)
Physical Form Crystalline
Quantity 1 g
UN Number UN1477
Solubility Information Decomposes in water; Soluble in HNO3
Formula Weight 445.99 (Anhydrous)
Percent Purity 99.99%
Grade Premion
Concentration or Composition (by Analyte or Components) Au 38.9 to 40.1%
Assay (metals basis)
Chemical Name or Material Hydrogen tetranitratoaurate(III) hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.