missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen tetrachloroaurate(III), solution, Au 38-42% w/w (cont. Au)

Product Code. 11400870
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11400870 1 g 1g
11410870 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11400870 Supplier Thermo Scientific Alfa Aesar Supplier No. 044744.NH
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16903-35-8 | HAuCl4

Hydrogen tetrachloroaurate(III) acts as a precursor used in the purification of gold by electrolysis. It is also used for the recovery, concentration, purification and analytical determination of gold. It acts as a key starting material for the preparation of gold compounds. It plays an important role as a catalyst in the preparation of 2-arylbenzoxazoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16903-35-8
Molecular Formula HAuCl4
MDL Number MFCD00011322

Specifications

Color Orange
UN Number UN3264
Concentration Au 40 to 44% w/w (cont. Au)
Solubility Information Miscible with water,alcohol,ester,ether and ketone.
Formula Weight 339.79
Quantity 1 g
Sensitivity Light sensitive
Physical Form Liquid
Chemical Name or Material Hydrogen tetrachloroaurate (III) solution

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.