missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen tetrachloroaurate(III) hydrate, Premion™, 99.995% (metals basis), Au 49% min

Product Code. 11335627
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11335627 5 g 5g
11345627 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11335627 Supplier Thermo Scientific Alfa Aesar Supplier No. 042803.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 27988-77-8 | AuCl4H | 339.77 g/mol

Hydrogen tetrachloroaurate(III) hydrate is used for microanalysis of rubidium, cesium and determination of alkaloids. Usually used as a harbinger for the preparation of gold (Au) nanoparticles. It is used in control rods in nuclear reactors and in the manufacture of light electric light filaments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 27988-77-8
Molecular Formula AuCl4H
Molecular Weight (g/mol) 339.77
MDL Number MFCD00149903
InChI Key RFWCEGCWRSNAGT-UHFFFAOYSA-K
IUPAC Name hydrogen tetrachlorogoldtris(ylium)
SMILES [H+].Cl[Au+3](Cl)(Cl)Cl

Especificaciones

Physical Form Crystalline
Quantity 5 g
Assay Percent Range 99.999% (metals basis), Au 49% min
Solubility Information Soluble in water. Insoluble in ether.
Formula Weight 339.79 (anhydrous)
Sensitivity Hygroscopic; Light sensitive
Chemical Name or Material Hydrogen tetrachloroaurate(III) hydrate

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.