missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen tetrachloroaurate(III) hydrate, 99.9% (metals basis), Au 49% min, Thermo Scientific Chemicals

Catalog Number p-7076341
Click to view available options
Quantity:
1 g
5 g
25 g
27988-77-8
MFCD00149903
hydrogen tetrachlorogoldtris(ylium)
[H+].Cl[Au+3](Cl)(Cl)Cl
27988-77-8
MFCD00149903
hydrogen tetrachlorogoldtris(ylium)
[H+].Cl[Au+3](Cl)(Cl)Cl

Used for microanalysis of rubidium, cesium and determination of alkaloids. Usually used as a harbinger for the preparation of gold (Au) nanoparticles. It is used in control rods in nuclear reactors and in the manufacture of light electric light filaments.

Hydrogen tetrachloroaurate(III) hydrate is used for microanalysis of rubidium, cesium and determination of alkaloids. Usually used as a harbinger for the preparation of gold (Au) nanoparticles. It is used in control rods in nuclear reactors and in the manufacture of light electric light filaments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Packaging Glass bottle
Molecular Formula AuCl4H
Quantity 1 g
UN Number UN3260
Sensitivity Hygroscopic; Light sensitive
Synonym Tetrachloroauric(III) acid,Chloroauric acid
Solubility Information Soluble in water. Insoluble in ether.
InChI Key RFWCEGCWRSNAGT-UHFFFAOYSA-K
Molecular Weight (g/mol) 339.77
Formula Weight 339.79 (anhydrous)
Percent Purity 99.9%
Physical Form Crystalline
Chemical Name or Material Hydrogen tetrachloroaurate(III) hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'