missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen tetrabromoaurate(III) hydrate, Premion™, 99.99% (metals basis), Au 32% min

Product Code. p-7051090
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11344038

Brand: Thermo Scientific Alfa Aesar 040433.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Au{C(NHMe) 2 } 2 ]Cl·H 2 O and [Au{C(NHMe) 2 } 2 ]Br·H 2 O were obtained by adding methyl isocyanide and 40% aqueous methylamine successively to aqueous solutions of hydrogen tetrachloroaurate(III) hydrate and hydrogen tetrabromoaurate(III) hydrate, respectively. To analyze the influence of the bromide ions in the synthesis of the gold nanorods Hydrogen tetrabromoaurate(III) hydrate is used. Hydrogen tetrabromoaurate(III) hydrate was used for purification in continuous microfluidic synthesis and functionalization of gold nanorods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00054118
Synonym Bromoauric acid

Specifications

Melting Point ∼27°C
Physical Form Crystalline
Quantity 1 g
UN Number UN3260
Solubility Information Soluble in water and alcohol
Formula Weight 517.61 (Anhydrous)
Percent Purity 99.99%
Grade Premion
Concentration or Composition (by Analyte or Components) Au 32% min.
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Hydrogen tetrabromoaurate(III) hydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.