missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen Peroxide Solution, Honeywell™ Fluka™

Product Code. 15654760 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 15654760

Brand: Honeywell-Fluka 349887500ML

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

35 wt. % in H2O

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 7722-84-1
Molecular Formula H2O2
MDL Number MFCD00011333
UN Number UN2014
Beilstein 3587191
Concentration 35 wt. % in H2O
Density 1.13g/mL at 25°C
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
SMILES OO
IUPAC Name hydrogen peroxide
Molecular Weight (g/mol) 34.014
PubChem CID 784
ChEBI CHEBI:16240
Formula Weight 34.01g/mol
Packaging Poly bottle
Vapor Pressure 23.3mmHg (30°C)
Quantity 500 mL
Chemical Name or Material Hydrogen peroxide solution
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Hydrogen peroxide solution
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : hydrogen peroxide solution
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.