missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen peroxide solution, Honeywell™

Product Code. 16656173 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
5 L
Unit Size:
5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16656173 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16656173 Supplier Honeywell Research Chemicals Supplier No. 40267H5L
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Semiconductor grade (Honeywell 17948), ≥30%

  • comprehensive portfolio for the electronics and semiconductor industry
  • for scientists engaged in education, research, development, prototyping and small scale fabrication of high tech micro- and nanochips
  • Superior purity, stability and exceptional lot-to-lot consistency
  • includes high purity products required for high performance applications such as etchants, solvents or hexamethyldisilazane (HMDS)
TRUSTED_SUSTAINABILITY

Specifications

CAS 7722-84-1, 7732-18-5
Boiling Point Decomposes on heating.
CAS Min % 68.90%, 30.00%
CAS Max % 69.90%, 31.00%
Molecular Formula H2O2
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
SMILES OO
IUPAC Name peroxol
Molecular Weight (g/mol) 34.01
PubChem CID 784
ChEBI CHEBI:16240
Packaging Plastic Bottle
Quantity 5 L
Physical Form Liquid
Grade Semiconductor grade reagent
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Hydrogen peroxide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: hydrogen peroxide solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.