missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen peroxide solution, Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
2.5 L
Unit Size:
2.50 L
Description
- Leading the industry for over 65 years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity and accuracy assured
Specifications
Specifications
| Color | Colorless |
| CAS | 7722-84-1 |
| Molecular Formula | H2O2 |
| InChI Key | MHAJPDPJQMAIIY-UHFFFAOYSA-N |
| SMILES | OO |
| IUPAC Name | peroxol |
| Molecular Weight (g/mol) | 34.01 |
| Grade | Synthesis |
| Packaging | Plastic Bottle |
| Quantity | 2.5 L |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction