missing translation for 'onlineSavingsMsg'
Learn More

Palintest™ Hydrogen Peroxide LR Test Kit

Product Code. 13121471
Click to view available options
Quantity:
1 Each
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 13121471

Brand: Palintest™ AP 104

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Concentration 0 to 2 mg/L
Quantity 1 Each
compliance-icons
Product Identifier
  • Test kit Palintest hydrogen peroxide LR re-fill
Signal Word
  • Danger
Hazard Category
  • Specific target organ toxicity Category 1
Hazard Statement
  • H372-Causes damage to organs.
  • EUH210-Safety data sheet available on request.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P314-Get medical advice/attention if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Potassium iodide, Molybdate, hexaammonium, tetrahydrate, Hexanedioic acid, Boronoxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.