missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen peroxide 30%, BAKER ANALYZED™ ACS, J.T. Baker™

Product Code. 15587984 Shop All J.T.Baker Products
Click to view available options
Quantity:
250 mL
2.5 L
Unit Size:
2.5 L
250mL
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 15587984

Brand: JT Baker 2192.0250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Exceeds ACS Specifications

  • Trace Impurities (in ppm):
    • Silver (Ag) max. 0.01
    • aluminum (Al) max. 0.05
    • Arsenic (As) max. 0.05
    • Barium (Ba) max. 0.02
    • Beryllium (Be) max. 0.01
    • Calcium (Ca) max. 0.5
    • Cadmium (Cd) max. 0.02
    • Cobalt (Co) max. 0.01
    • Chromium (Cr) max. 0.1
    • Copper (Cu) max. 0.01
    • Iron (Fe) max. 0.1
    • Germanium (Ge) max. 0.05
    • Potassium (K) max. 0.1
    • Lithium (Li) max. 0.01
    • Magnesium (Mg) max. 0.1
    • Manganese (Mn) max. 0.01
    • Molybdenum (Mo) max. 0.02
    • Sodium (Na) max. 0.5
    • Nickel (Ni) max. 0.05
    • Lead (Pb) max. 0.05
    • Strontium (Sr) max. 0.01
    • Titanium (Ti) max. 0.1
    • Thallium (Tl) max. 0.05
    • Vanadium (V) max. 0.01
    • Zinc (Zn) max. 0.05
    • Zirconium (Zr) max. 0.1

Chemical Identifiers

CAS 7722-84-1
Molecular Formula H2O2
Molecular Weight (g/mol) 34.01
MDL Number MFCD00011333
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
IUPAC Name peroxol
SMILES OO

Specifications

Color Max. 10 (APHA)
Boiling Point ∼107°C (1013 hPa)
Melting Point ≤0°C
Assay Percent Range 30.0 to 32.0%
Density 1.11 g/cm³ (20°C)
Packaging Plastic Bottle
Residue after Evaporation max. 0.002%
Quantity 250 mL
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Oxidising liquid Category 2
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : hydrogen peroxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.