missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen Peroxide 30-32% (w/w) (100 Volumes), Certified AR for Analysis, Fisher Chemical™

Product Code. 11488333 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
HDPE plastic bottle
Plastic container
Unit Size:
1 L
2.5 L
25 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11488333 25 L Plastic container 25 L
10736291 1 L HDPE plastic bottle 1 L
10284000 2.5 L HDPE plastic bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 11488333 Supplier Fisher Chemical Supplier No. H/1800/25
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

H2O2, CAS Number-7722-84-1, albone, hioxyl, hydrogen dioxide, inhibine, interox, kastone, oxydol, perhydrol, peroxaan, superoxol, 1L, 30-32%w/w, 108 deg.C, CHEBI:16240, Colorless, MHAJPDPJQMAIIY-UHFFFAOYSA-N, hydrogen peroxide, -33 deg.C, 11333, 34.014, HDPE plastic bottle, 3.3, Liquid, 784, OO, 1.1

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7722-84-1
Molecular Formula H2O2
Molecular Weight (g/mol) 34.014
MDL Number 11333
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name hydrogen peroxide
SMILES OO

Specifications

Color Colorless
Boiling Point 108°C
Melting Point -33°C
Assay Percent Range 30-32%w/w
Packaging Plastic container
Vapor Density 1.1
pH 3.3
Quantity 25 L
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.