missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzonitrile, 98+%

Product Code. 11377976
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11377976 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 11377976 Supplier Thermo Scientific Alfa Aesar Supplier No. L13235.AE
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

CAS: 7722-84-1 | H2O2 | 34.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7722-84-1
Boiling Point 107°C
Melting Point -26°C
Molecular Formula H2O2
MDL Number MFCD00011333
UN Number UN2014
Beilstein 3587191
Concentration 27% w/w aq. soln.
Density 1.1
Merck Index 14,4798
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide
Solubility Information Miscible with water,ether,alcohol,organic solvents and carboxylic esters. Immiscible with hydrocarbons and petroleum ether.
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
SMILES OO
IUPAC Name peroxol
Molecular Weight (g/mol) 34.01
PubChem CID 784
ChEBI CHEBI:16240
Formula Weight 34.02
Refractive Index 1.335
Quantity 100 mL
Chemical Name or Material Hydrogen peroxide, stabilized
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.