missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen peroxide, 12% w/v

Product Code. 16534274 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
Unit Size:
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16534274 100 mL 100mL
16524274 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16534274 Supplier Thermo Scientific Acros Supplier No. 464571000
  • ONLINE EXCLUSIVE PRICE
    £15.00
    £13.65 / 100mL
    Save £1.35
    9% Off
View all promotions
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 7722-84-1 | H2O2 | 34.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7722-84-1
Molecular Weight (g/mol) 34.01

Specifications

Boiling Point 100.0°C
Density 1.0500 g/mL
Quantity 100 mL
Molecular Formula H2O2
Linear Formula H2O2
Specific Gravity 1.05
Solubility Information Solubility aq. soln.: soluble.
Formula Weight 34.01
Chemical Name or Material Hydrogen peroxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.