missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen hexachloroplatinate hydrate, 99.995%, (trace metal basis)

Product Code. 10506440 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10506440 1 g Glass bottle 1g
10441990 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10506440 Supplier Thermo Scientific Acros Supplier No. 437200010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 26023-84-7 | Cl6H2Pt | 409.80 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26023-84-7
Molecular Formula Cl6H2Pt
Molecular Weight (g/mol) 409.80
MDL Number MFCD00149909
InChI Key ZKOQTCCVSRSECD-UHFFFAOYSA-J
Synonym Hexachloroplatinic acid hydrate, Platinic chloride hydrate
IUPAC Name dihydrogen hexachloroplatinumtetrakis(ylium)
SMILES [H+].[H+].Cl[Pt+4](Cl)(Cl)(Cl)(Cl)Cl

Specifications

Melting Point 60.0°C
Physical Form Powder, Crystals and/or Chunks
Quantity 1 g
Assay Percent Range 37% min. (Pt)
Linear Formula H2PtCl6·xH2O
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol
Formula Weight 409.82
Percent Purity 99.995%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Hydrogen hexachloroplatinate hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.