missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen hexachloroiridate(IV) hexahydrate, approx. 40% Iridium

Product Code. 10137503 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10137503 5 g Glass Bottle 5g
10022560 1 g Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10137503 Supplier Thermo Scientific Acros Supplier No. 197550050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16941-92-7 | H2Cl6Ir·6H2O | 515.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16941-92-7
Molecular Formula H2Cl6Ir·6H2O
Molecular Weight (g/mol) 515.02
MDL Number MFCD00149907
Synonym Chloroiridic acid

Specifications

Color Black
Packaging Glass Bottle
Linear Formula H2IrCl6·6H2O
Quantity 5 g
Fieser 01,131; 02,67; 03,47; 04,93; 05,119; 05,287; 06,45; 17,269; 05,203; 06,108; 07,59; 08,434; 10,382; 14,294; 17,326
Solubility Information Solubility in water: soluble
Formula Weight 515.02
Percent Purity ∼40% Iridium
Physical Form Crystals
Chemical Name or Material Hydrogen hexachloroiridate(IV) hexahydrate, Approx. 40% Iridium

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.