Learn More
Hydrogen hexachloroiridate(IV) hexahydrate, approx. 40% Iridium, ACROS Organics™
Brand: Acros Organics 197550010
Code : 29
Additional Details : CAS Number : 16941-92-7 Weight : 0.05100kg Transport : UN number : 3260 Chem class : 8 Pack group : III
Packaging | Glass Bottle |
---|---|
Quantity | 1g |
Chemical Identifiers
16941-92-7 | |
515.02 | |
Chloroiridic acid |
H2Cl6Ir·6H2O | |
MFCD00149907 |
Specifications
Hydrogen hexachloroiridate(IV) hexahydrate | |
Black | |
Approx. 40% Iridium | |
H2Cl6Ir·6H2O | |
MFCD00149907 | |
Chloroiridic acid | |
515.02 | |
Crystals |
Glass Bottle | |
1g | |
16941-92-7 | |
H2IrCl6·6H2O | |
01,131; 02,67; 03,47; 04,93; 05,119; 05,287; 06,45; 17,269; 05,203; 06,108; 07,59; 08,434; 10,382; 14,294; 17,326 | |
Solubility in water: soluble | |
515.02 | |
approx. 40% Iridium |