missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen fluoride pyridine complex, ca 70% HF, Thermo Scientific Chemicals

Product Code. p-7046996
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11373956

Brand: Thermo Scientific Alfa Aesar L17117.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hydrogen fluoride pyridine complex is used to prepare beta-fluoroamines via reaction with amino alcohols. As a fluorinating agent, is involved in the fluorination of 2,2-diary-1,3-dithiolanes. It is associated with nitrosonium tetrafluorborate to prepare gem-difluorides by reacting with ketoximes. It is also used to convert secondary and tertiary alcohols into their respective alkyl fluorides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hydrogen fluoride pyridine complex is used to prepare beta-fluoroamines via reaction with amino alcohols. As a fluorinating agent, is involved in the fluorination of 2,2-diary-1,3-dithiolanes. It is associated with nitrosonium tetrafluorborate to prepare gem-difluorides by reacting with ketoximes. It is also used to convert secondary and tertiary alcohols into their respective alkyl fluorides.

Notes
Moisture sensitive. Store in cool place. Incompatible with strong bases, alkali metals, strong oxidizing agents, metals and strong acids. It reacts violently with water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62778-11-4
Molecular Formula C5H6FN
Molecular Weight (g/mol) 99.108
MDL Number MFCD00012436
InChI Key GRJJQCWNZGRKAU-UHFFFAOYSA-N
Synonym pyridine hydrofluoride, hydrogen fluoride-pyridine, olah's reagent, pyridinium fluoride, hydrogen fluoride pyridine complex, hydrogen fluoride pyridine, hf-pyridine, pyridine, hydrofluoride 1:1, pyridinium poly hydrogen fluoride, hydrogen fluoride/pyridine
PubChem CID 64774
IUPAC Name pyridine;hydrofluoride
SMILES C1=CC=NC=C1.F

Specifications

Density 1.1
Boiling Point 50°C (1 mmHg)
Quantity 25 g
UN Number UN1790
Beilstein 4750151
Sensitivity Moisture sensitive
Merck Index 14,6821
Assay ≈70% HF
Chemical Name or Material Hydrogen fluoride pyridine complex
compliance-icons compliance-icons
Product Identifier
  • Hydrogen fluoride pyridine complex
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
Supplemental information
  • MIXTURE LIST-Contains : Hydrogen fluoride pyridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.