missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen Chloride Solution, 2M in Diethyl Ether, Honeywell™

Product Code. 15675560 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
Glass Bottle
Unit Size:
100mL
800mL
This item is not returnable. View return policy

Product Code. 15675560

Brand: Honeywell-Fluka 455180100ML

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Color Colorless
Boiling Point 34.6°C
UN Number UN2924
Density 0.747g/mL at 25°C
Formula Weight 36.46g/mol
Packaging Glass Bottle
Quantity 100 mL
Flash Point −34°C (−29.2°F)
Physical Form Liquid
Chemical Name or Material Hydrogen chloride solution
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.