missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen chloride, pure, 5 to 6N solution in 2-propanol

Product Code. 11613069 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Plastic Bottle
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11613069 2.5 L Plastic Bottle 2.5 L
10214950 1 L Plastic Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price
Product Code. 11613069 Supplier Thermo Scientific Acros Supplier No. 133700025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27968” to get your promotional price

CAS: 7647-01-0 | ClH | 36.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hydrochloric acid
CAS 7647-01-0, 67-63-0
Color Colorless to Yellow
Physical Form Liquid
Quantity 2.5 L
Molecular Formula ClH
Linear Formula HCl
MDL Number MFCD00011324 MFCD00792839
Fieser 04,450; 05,533; 06,283
Flash Point 11°C
Merck Index 15,4831
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
Solubility Information Solubility in water: >1000g/L (20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol: >50g/L
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES Cl
IUPAC Name hydrogen chloride
Molecular Weight (g/mol) 36.46
PubChem CID 313
ChEBI CHEBI:17883
Formula Weight 36.45
Grade Pure
Concentration or Composition (by Analyte or Components) 5N min.
Packaging Plastic Bottle
Specific Gravity 0.909
Density 0.9090g/mL
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.