missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen chloride, 4N solution in 1,4-Dioxane, standard cap, Thermo Scientific Chemicals

Product Code. 15731819 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15731819 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15731819 Supplier Thermo Scientific Acros Supplier No. 455040010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7647-01-0
Density 1.0500 g/mL
Specific Gravity 1.05
Flash Point 17°C
Quantity 1 L
Molecular Formula ClH
Linear Formula HCl
Merck Index 15, 4831
Solubility Information Solubility aq. soln.: soluble. Other solubilities: soluble in most organic solvents
Molecular Weight (g/mol) 36.45
Formula Weight 36.45
Chemical Name or Material Hydrochloric acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.