missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen chloride, 1M in acetic acid

Product Code. p-7029835
Click to view available options
Quantity:
100 mL
1000 mL
Unit Size:
1000mL
100mL
This item is not returnable. View return policy

Product Code. 11327099

Brand: Thermo Scientific Alfa Aesar H27347.AE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7647-01-0 | ClH | 36.46 g/mol

In chemical industry production of inorganic salts, Production of vinyl chloride from acetylene, alkyl chlorides from olefins, dyes, artificial silk, pigment, s, pharmaceutical hydrochloride, As catalyst and solvent in organic syntheses, Alcohol chlorination reagent, Nitration reaction, Manufacture of phosphoric acid and in the production of ammonium chloride, Neutralization of basic systems, Cleaning agents. Hydrolyzing of starch and proteins in the preparation of various food products. The hydrogen chloride gas is mainly used for EDC and vinyl chloride monomer (VCM) production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
SMILES Cl

Specifications

UN Number UN2920
Concentration 1M in acetic acid
Density 1.05
Solubility Information Miscible in water.
IUPAC Name hydrogen chloride
Formula Weight 36.46
Odor Acrid
Quantity 100 mL
Flash Point 40°C (104°F)
Sensitivity Hygroscopic
Chemical Name or Material Hydrochloric acid
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hydrogen chloride, 1M in acetic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Acetic acid, Hydrochloric acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.