missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen bromide solution, 33 wt. % in acetic acid, Honeywell Fluka™

Product Code. 15602710 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 mL
500 mL
2 L
Unit Size:
100mL
2 L
500mL
This item is not returnable. View return policy

Product Code. 15602710

Brand: Honeywell-Fluka 2486302L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

33 wt. % in acetic acid

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10035-10-6, 64-19-7
Molecular Formula BrH
Molecular Weight (g/mol) 80.91
MDL Number MFCD00011323
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-N
Synonym hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
PubChem CID 260
ChEBI CHEBI:47266
SMILES Br

Specifications

CAS Min % ≥50.0000%, ≥30.0000%
CAS Max % <70.0000%, <50.0000%
Linear Formula HBr
UN Number UN3265
Density 1.354 g/mL at 25 °C
IUPAC Name hydrogen bromide
Grade Acids/Bases
Quantity 2 L
Flash Point 65°C
Chemical Name or Material Hydrogen bromide solution, 33 wt.% in acetic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.