missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen bromide, pure, 33 wt% solution in glacial acetic acid
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass Bottle
Unit Size:
1 L
2.5 L
500mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
Specifications
Specifications
| Color | Yellow |
| Physical Form | Fuming Liquid |
| Quantity | 1 L |
| Linear Formula | HBr |
| UN Number | 2922 |
| Fieser | 01,450; 02,214; 03,154; 04,249; 05,332; 06,282; 10,200; 15,273 |
| Flash Point | 39°C |
| Merck Index | 09, 4665 |
| IUPAC Name | bromane |
| Formula Weight | 80.9 |
| Show More |
Safety and Handling
Product Identifier
- Hydrogen bromide, 33 wt% solution in glacial acetic acid
- Danger
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 3
- H226-Flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Acetic acid, Hydrogen bromide
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction