missing translation for 'onlineSavingsMsg'
Learn More

Hydrofluoric Acid, TRC

Product Code. 30406833
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
30406833 1 g 1g
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30406833

Brand: Toronto Research Chemicals TRCH9424501G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hydrofluoric Acid
CAS 7664-39-3
Molecular Formula FH
Quantity 1 g
Synonym Alsurf 45, Anhydrous hydrofluoric Acid, Antisal 2b, Fluorhydric Acid, Fluoric Acid, Fluorine Hydride (FH), Fluorine Monohydride, G-C Hydrofluoride Gel, Hydrofluoric Acid Gas, Hydrogen Fluoride, Hydrogen Fluoride (HF), Hydrogen Monofluoride, Porcelain Conditioner, SA-X, Ultradent Porcelain Etch, Urine Luck 6.3, Vita Ceramic etch
InChI Formula InChI=1S/FH/h1H
SMILES F
IUPAC Name fluorane
Molecular Weight (g/mol) 20.0064
Formula Weight 20.0062
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.