Product Code. 15610590
Shop All Honeywell Fluka Products
Click to view available options
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
Unit Size
15610590
250 mL
250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code.
15610590
Supplier
Honeywell-Fluka
Supplier No.
02658250ML
This item is currently unavailable or has been discontinued. View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Specifications
CAS
7664-39-3
Concentration
≥49%
Density
1.15g/mL at 25°C
Vapor Pressure
25mmHg (20°C)
Quantity
250 mL
Molecular Formula
FH
MDL Number
MFCD00011346
UN Number
1790
Synonym
hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484
InChI Key
KRHYYFGTRYWZRS-UHFFFAOYSA-N
SMILES
F
IUPAC Name
hydrogen fluoride
Molecular Weight (g/mol)
20.01
PubChem CID
14917
ChEBI
CHEBI:29228
Formula Weight
20.01g/mol
Grade
TraceSELECT™ Ultra
Chemical Name or Material
Hydrofluoric Acid
Show More
Show Less
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?