missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrofluoric Acid 47-51%, Optima™, for Ultra Trace Elemental Analysis, Fisher Chemical™
£490.00 - £2050.00
Chemical Identifiers
CAS | 7664-39-3 |
---|---|
Molecular Formula | FH |
Molecular Weight (g/mol) | 20.01 |
MDL Number | MFCD00011346 |
InChI Key | KRHYYFGTRYWZRS-UHFFFAOYSA-N |
IUPAC Name | hydrogen fluoride |
SMILES | F |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11944101
|
Fisher Chemical
A463-250 |
250 mL |
£490.00
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11954101
|
Fisher Chemical
A463-500 |
500 mL |
£962.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11934101
|
Fisher Chemical
A463-1 |
1 L | PFA Bottle |
£2050.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
7664-39-3 | |
20.01 | |
KRHYYFGTRYWZRS-UHFFFAOYSA-N | |
F |
FH | |
MFCD00011346 | |
hydrogen fluoride |
Specifications
1 | |
7664-39-3 | |
105°C | |
Liquid | |
KRHYYFGTRYWZRS-UHFFFAOYSA-N | |
hydrogen fluoride | |
20g/mol |
Colorless | |
-35°C | |
FH | |
MFCD00011346 | |
F | |
20.01 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Hydrofluoric acid 47-51%, Optima for Ultra traces analysis
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Acute toxicity Category 2
- Acute toxicity Category 1
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H290-May be corrosive to metals.
- H300-Fatal if swallowed.
- H310-Fatal in contact with skin.
- H330-Fatal if inhaled.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P262-Do not get in eyes, on skin, or on clothing.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.