missing translation for 'onlineSavingsMsg'
Learn More

Hydrofluoric acid, J.T. Baker™

Product Code. 15508644 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
POLYETHYLENE BOTTLE
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15508644 500 mL POLYETHYLENE BOTTLE Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15508644 Supplier Avantor J.T.Baker Supplier No. 956001

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Color Colorless
CAS 7664-39-3
Melting Point -83.6°C
Boiling Point 19.5°C
Density 1.15g/mL
Packaging POLYETHYLENE BOTTLE
Specific Gravity 1L = 1.18kg
Quantity 500 mL
Molecular Formula FH
Physical Form Liquid
MDL Number MFCD00011346
Synonym hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484
InChI Key KRHYYFGTRYWZRS-UHFFFAOYSA-N
SMILES F
IUPAC Name fluorane
Molecular Weight (g/mol) 20.01
PubChem CID 14917
ChEBI CHEBI:29228
Formula Weight 20.01
Percent Purity 48.0 to 51.0%
Grade BAKER ANALYZED™ ACS Reagent
Chemical Name or Material Hydrofluoric acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.