missing translation for 'onlineSavingsMsg'
Learn More

Hydrofluoric acid, Honeywell

Product Code. 15694100 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15694100 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15694100 Supplier Honeywell Research Chemicals Supplier No. 339261100ML

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

48 wt. % in H2O,≥99.99% trace metals basis

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Specifications

CAS 7664-39-3
Concentration 48 wt. % in H2O
Density 1.15g/mL at 25°C
Vapor Pressure 25 mmHg (20°C)
Quantity 100 mL
Molecular Formula FH
MDL Number MFCD00011346
UN Number UN1790B
Synonym hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484
InChI Key KRHYYFGTRYWZRS-UHFFFAOYSA-N
SMILES F
IUPAC Name hydrogen fluoride
Molecular Weight (g/mol) 20.01
PubChem CID 14917
ChEBI CHEBI:29228
Formula Weight 20.01g/mol
Percent Purity ≥99.99% (trace metals basis)
Chemical Name or Material Hydrofluoric acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.